Learn More
Enterprise Technology Solutions MCR Safety Safety Glasses, Wraparound, Clear/Chrome
SDP
Catalog No. 5000828037
Encompass_Preferred
Safety glasses feature durable, metal alloy temples with nonslip temple sleeves for a comfortable, secure fit. Scratch-resistant coating ensures high visibility and long-lasting use. Rimless design offers excellent unobstructed vision. Clear, polycarbonate lens filters 99.9 percent of UV rays.
Catalog No. 50-008-28037 Supplier Enterprise Technology Solutions Supplier No. MCSCRWS2110
Safety glasses feature durable, metal alloy temples with nonslip temple sleeves for a comfortable, secure fit. Scratch-resistant coating ensures high visibility and long-lasting use. Rimless design offers excellent unobstructed vision. Clear, polycarbonate lens filters 99.9 percent of UV rays.