Learn More
Enterprise Technology Solutions MCR Safety Safety Goggle, w/ Adjustable Strap, Antifog, Clear
SDP
Catalog No. 5000828362
Encompass_Preferred
Goggles offer a clear anti-fog protection from chemical splashes while providing you with ventilation. Rubber strap offers a secure fit. Goggle surface with Duramass AF4 anti-fog coating offers superior resistance to scratches and helps prevent fogging. Goggles meet or exceed ANSI Z87.1 safety standards.
Catalog No. 50-008-28362 Supplier Enterprise Technology Solutions Supplier No. MCS2310AF
Goggles offer a clear anti-fog protection from chemical splashes while providing you with ventilation. Rubber strap offers a secure fit. Goggle surface with Duramass AF4 anti-fog coating offers superior resistance to scratches and helps prevent fogging. Goggles meet or exceed ANSI Z87.1 safety standards.