Learn More
Enterprise Technology Solutions MCR Safety Safety Goggles, Indirect Vent, Adj Strap, Clear
SDP
Catalog No. 5000828216
Encompass_Preferred
Goggles provide a wide, unobstructed field of vision. Large design can be worn with most half-mask respirators. The transparent, protective PVC body permits a comfortable fit over prescription eyewear while protecting against impact, flying particles and debris.
Catalog No. 50-008-28216 Supplier Enterprise Technology Solutions Supplier No. MCSCRW2230R
Goggles provide a wide, unobstructed field of vision. Large design can be worn with most half-mask respirators. The transparent, protective PVC body permits a comfortable fit over prescription eyewear while protecting against impact, flying particles and debris.