Learn More
MSC MCR Safety Safety Vest, ANSI/ISEA 107 Class 3, 1 ea
Catalog No. 5000766241
Encompass
MCR Safety has expanded its high visibility line to include a flame resistant safety vest made with GlenGuard® mesh lime modacrylic / aramid blended fabric with FR reflective stripes. The vest features three pockets, a D-Ring access slot on the back, durable gray binding, and a hook and loop front closure. The ANSI Class 3 vest also meets ASTM F1506 and conforms to NFPA 70E- HRC1 specifications with an Arc Rating of 4.6 cal/cm².
  • Two inch silver FR reflective stripes
  • Three pockets
  • ANSI/ISEA 107 Type R, Class 3 compliant
  • Arc rating 4.6 cal/cm²
  • Conforms to NFPA 70E, HRC1
  • D-ring access for fall protection harness
  • Hook and loop front closure
  • Mesh modacrylic/aramid blend fabric
  • Meets ASTM F1506
Catalog No. 50-007-66241 Supplier MSC Supplier No. 94632585
MCR Safety has expanded its high visibility line to include a flame resistant safety vest made with GlenGuard® mesh lime modacrylic / aramid blended fabric with FR reflective stripes. The vest features three pockets, a D-Ring access slot on the back, durable gray binding, and a hook and loop front closure. The ANSI Class 3 vest also meets ASTM F1506 and conforms to NFPA 70E- HRC1 specifications with an Arc Rating of 4.6 cal/cm².
  • Two inch silver FR reflective stripes
  • Three pockets
  • ANSI/ISEA 107 Type R, Class 3 compliant
  • Arc rating 4.6 cal/cm²
  • Conforms to NFPA 70E, HRC1
  • D-ring access for fall protection harness
  • Hook and loop front closure
  • Mesh modacrylic/aramid blend fabric
  • Meets ASTM F1506