Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Select Shoulder Split Cowhide Leather Welding Work Gloves

Catalog No. 19083542 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
ANSI Abrasion Rating:
ANSI 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. ANSI Abrasion Rating
19-083542 ANSI 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-083542 Supplier MCR Safety Supplier No. 4150
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from select shoulder split cowhide leather with a one piece back and cotton drill lining, ideal for use with welding applications.

  • Full cotton sock lining
  • One piece back
  • Welted seams
  • Natural pearl color
  • Generous sizing

Specifications

ANSI Abrasion Rating ANSI 4
Color Gray
Grade Select
Hand Specific
Material Shoulder Split Cowhide Leather
Size X-Large
Style Gunn
Glove Cuff Style Gauntlet
Hem Color Brown
Liner Material Fleece
Packaging Type Polybag
Series 4150
Thumb Style Wing
Length (English) 13.6 in.
Length (English) Cuff 5.3 in.
Length (Metric) 34.5 cm
Length (Metric) Cuff 13.3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.