Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Steelcore™ II Cut-Resistant Gloves

Catalog No. 191600044 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-160-0044 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-160-0044 Supplier MCR Safety Supplier No. 9350L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Standard weight cut-resistant gloves are made of strands of stainless steel wrapped with polyester fiber to add strength and comfort.

  • Made from dual stainless steel wrapped with softer polyester yarn
  • Launderable without shrinkage
  • Reversible
  • 7-gauge
  • Color: yellow

Canning, food processing, glass handling, metal fabrication

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance FDA accepted material for use in food processing
Construction String Knit
Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A8
Hand Ambidextrous
Latex Free No
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) food processing
Glove Cuff Style Knit
Hem Color Yellow
Model 9350L
Series Steelcore™ II
Yarn Gauge 7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.