Learn More
MCR Safety Storm™Safety Glasses
Catalog No. 19046160 Shop All MCR Safety Products
Change view
Click to view available options
Lens Tint:
Green 3.0
Green 5.0
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-046-160 Green 3.0
19-046-161 Green 5.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-046-160 Supplier MCR Safety Supplier No. ST1130
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Workers in the Storm™ can expect extreme comfort with this face-flattering protective eyewear from Crews™. Ratchet action temples combined with non- slip rubber head grips create a customized fit for virtually any head size.
Wraparound lens constructed of lightweight, scratch resistant polycarbonate, that filters 99.9% of U.V. rays, delivers advanced visual acuity and protection.
  • Available in a variety of frame colors and lens combinations
  • Single wraparound dual focal point lens with integral side shields
  • 5 position ratchet action bayonet temples
  • Exclusive Duramass™ scratch-resistant coating
  • Non-slip, soft temple material
Specifications
Lens Coating Scratch-resistant
Lens Tint Green 3.0
Product Type Safety Glasses
Frame Color Black
Frame Material Nylon