Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Crinkle Latex Coated Cotton Jersey Work Gloves

Catalog No. 19041917 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Coated Area:
Palm, Fingers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coated Area
19-041917 Palm, Fingers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-041917 Supplier MCR Safety Supplier No. 6830
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Palm, fingers, and thumb are coated with heavy crinkle-textured latex for durable gripping power. Knit wrist helps prevent dirt and debris from entering the glove.

  • Crinkle rubber latex coated palm, fingers, and thumb
  • Knit wrist helps keep dirt and debris from entering the glove
  • Jersey lining offers lightweight and durable comfort

Specifications

Coated Area Palm, Fingers
Coating Material Crinkled Latex
Color Yellow
Exterior Finish Fully Textured
Grade Industrial
Hand Specific
Latex Free No
Material Cotton
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Automotive Assembly, Landscaping, Construction, and Manufacturing
Glove Cuff Color Natural
Glove Cuff Style Knit
Series 6830
Thumb Style Wing
Length (English) 10 in.
Length (Metric) 25.4 cm
Show More Show Less

WARNING: Cancer and Reproductive Harm www.P65Warnings.ca.gov/product

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.