Learn More
MCR Safety Cut Pro™ 9676 Dyneema™ Cut-Resistant Gloves, Polyurethane Coating
Shop All MCR Safety Products
Description
- 13-gauge salt and pepper colored DSM Certified Dyneema™ shell
- Cool and comfortable
- Cut-resistant, abrasion resistant, exceptional dexterity
- Gray polyurethane coated palm and fingertips
- Treated with ActiFresh™
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 6 |
| Coating Material | Polyurethane |
| Color | Gray |
| Cut Resistance Rating | ANSI A2 |
| Material | Dyneema |
| Product Type | Cut-resistant Gloves |
| Size | X-Large |
| For Use With (Application) | Abrasive applications, assembly, brick handling, construction, farming, general maintenance, landscaping, plumbing, recycling, shipping/receiving |
| Glove Cuff Color | Green |
| Glove Cuff Style | Knit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.