Learn More
MSC MCR Safety Welding Gloves, Pigskin Leather, Medium, 1 Pair
Catalog No. 5000684234
Encompass
The MCR Safety 49600 welding gloves are crafted from premium A grade black grain pigskin, featuring a 4.5-inch split cow cuff. These gloves are stitched with DuPont™ Kevlar® thread for resistance against sparks and sharp metals, making them ideal for low-heat welding applications requiring optimal dexterity and detailed work.
  • Premium A grade top grain pigskin leather
  • Gray, 4.5-inch split cowhide leather gauntlet cuff
  • Sewn with super-strong, heat-resistant DuPont™ Kevlar® thread
  • Wing thumb design
  • Unlined for optimal dexterity
  • Clute back pattern
Catalog No. 50-006-84234 Supplier MSC Supplier No. 66281650
The MCR Safety 49600 welding gloves are crafted from premium A grade black grain pigskin, featuring a 4.5-inch split cow cuff. These gloves are stitched with DuPont™ Kevlar® thread for resistance against sparks and sharp metals, making them ideal for low-heat welding applications requiring optimal dexterity and detailed work.
  • Premium A grade top grain pigskin leather
  • Gray, 4.5-inch split cowhide leather gauntlet cuff
  • Sewn with super-strong, heat-resistant DuPont™ Kevlar® thread
  • Wing thumb design
  • Unlined for optimal dexterity
  • Clute back pattern