Learn More
MSC MCR Safety Welding Jacket, Limited Flammability, Medium, 1 ea
Catalog No. 5000675228
Encompass
This green cotton welding jacket provides economical protection from light molten splashes and sparks. Made from 9-oz. cotton, it is comfortable for extended wear and easily laundered.
  • Made from 100% 9-oz. green finish cotton
  • Anodized snap closures resist corrosion and wear
  • Length is 30 in. for ample coverage
  • Limited flammability, meeting ASTM D 6413 and NFPA-701 standards
  • Lightweight fabric offers comfort
Catalog No. 50-006-75228 Supplier MSC Supplier No. 43828318
This green cotton welding jacket provides economical protection from light molten splashes and sparks. Made from 9-oz. cotton, it is comfortable for extended wear and easily laundered.
  • Made from 100% 9-oz. green finish cotton
  • Anodized snap closures resist corrosion and wear
  • Length is 30 in. for ample coverage
  • Limited flammability, meeting ASTM D 6413 and NFPA-701 standards
  • Lightweight fabric offers comfort