Learn More
MCR Safety™ Wizard PVC/Nylon Rainwear: Rain Coats
Supplier:  MCR Safety™ 300CX2
Catalog No. 19-088-1712E
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Double-coated PVC over 210 denier nylon scrim material
  • Rip-stop material provides excellent resistance to snags and tears
  • Protects against most chemicals, oils and acids
  • Features welded seams
  • 0.28mm PVC/Nylon/PVC construction
Raincoat
  • Snap-storm-fly front with take-up snaps on wrists
  • Snap-on, detachable drawstring hood
  • Two patch pockets with flap and closure
  • Raglan sleeves with underarm air vents
For applications like trucking, fertilizer plants, sanitation workers and paper mills
Product Suggestions
Product Suggestions