Learn More
MSC MCR Safety Work Gloves, Cowhide, Medium, Safety Cuff, 1 ea
Catalog No. 5000803362
Encompass
MCR Safety 1400 series leather palm work gloves provide protection against abrasion, punctures, and light scratches and cuts. Crafted from select B+ grade split shoulder leather, these durable gloves feature a 2.5 inch plasticized safety cuff and are designed for repeated laundering.
  • Constructed from select B+ grade split shoulder leather
  • Features a 2.5 inch plasticized safety cuff
  • Includes color coded hems for identifying sizing
  • Equipped with a leather knuckle strap
  • Wing thumb style for enhanced dexterity
  • Lined with fleece for added comfort
Catalog No. 50-008-03362 Supplier MSC Supplier No. 73059388
MCR Safety 1400 series leather palm work gloves provide protection against abrasion, punctures, and light scratches and cuts. Crafted from select B+ grade split shoulder leather, these durable gloves feature a 2.5 inch plasticized safety cuff and are designed for repeated laundering.
  • Constructed from select B+ grade split shoulder leather
  • Features a 2.5 inch plasticized safety cuff
  • Includes color coded hems for identifying sizing
  • Equipped with a leather knuckle strap
  • Wing thumb style for enhanced dexterity
  • Lined with fleece for added comfort