Ninja Ice Insulated Work Gloves feature a hi-vis 15 gauge lime nylon shell with a 7 gauge acrylic terry interior liner, providing warmth and high visibility. The HPT coated palm and fingertips repel liquids and maintain softness in cold temperatures, ensuring a firm grip in both wet and dry conditions for various applications.
- Acrylic terry liner for insulation and warmth
- Allows air to flow through, keeping hands dry
- Coating remains soft in temperatures as low as -4° F
- Encapsulated air molecules provide a cushioning effect to minimize impact
- HPT coating provides a 360 anti-wet treatment that keeps liquids away from the glove
- Over-the-knuckle coating for added protection
- Porous HPT coating repels liquid for an unprecedented firm grip in wet or dry conditions
- Treated with ActiFresh® to help control bacteria and odor