Learn More
Enterprise Technology Solutions MCR Safety Work Gloves,Industrial,FlatDip,Ergonomic,13GANylon,PU,XS,BK
SDP
Catalog No. 5000828253
Encompass_Preferred
9669 NXG Gloves deliver next-generation performance through advanced manufacturing and thoughtful design. Gloves feature seamless 13-gauge nylon shells and polyurethane palm coating that enhance grip and abrasion resistance without compromising dexterity. Flat-dipped coating and ergonomic liner mimic the natural curve of your hand to reduce fatigue and boost productivity. Gloves are produced in state-of-the-art facilities using environmentally responsible methods and come free of silicone.
Catalog No. 50-008-28253 Supplier Enterprise Technology Solutions Supplier No. MCSCRW9669XS
9669 NXG Gloves deliver next-generation performance through advanced manufacturing and thoughtful design. Gloves feature seamless 13-gauge nylon shells and polyurethane palm coating that enhance grip and abrasion resistance without compromising dexterity. Flat-dipped coating and ergonomic liner mimic the natural curve of your hand to reduce fatigue and boost productivity. Gloves are produced in state-of-the-art facilities using environmentally responsible methods and come free of silicone.