Learn More
MSC MCR Safety Work Gloves, Large, Latex-Coated, Acrylic, 1 Pair
Catalog No. 5000677786
Encompass
These general purpose work gloves are designed for various applications, offering protection against abrasion, heat, cut, and puncture. They feature a latex-coated palm and fingertips for enhanced grip, and an acrylic lining for comfort.
  • Acrylic material and lining
  • Latex coating on palm and fingertips with crinkle grip
  • Knit cuff style
  • Full-fingered design
  • High visibility lime and blue color
  • Provides protection against abrasion, heat, cut, and puncture
  • Unisex, large size (numeric size 9)
  • Maximum temperature of 500°F
Catalog No. 50-006-77786 Supplier MSC Supplier No. 02485316
These general purpose work gloves are designed for various applications, offering protection against abrasion, heat, cut, and puncture. They feature a latex-coated palm and fingertips for enhanced grip, and an acrylic lining for comfort.
  • Acrylic material and lining
  • Latex coating on palm and fingertips with crinkle grip
  • Knit cuff style
  • Full-fingered design
  • High visibility lime and blue color
  • Provides protection against abrasion, heat, cut, and puncture
  • Unisex, large size (numeric size 9)
  • Maximum temperature of 500°F