Learn More
MSC MCR Safety Work Gloves, Latex Coated, Crinkle Finish, 1 Pair
Catalog No. 5000792048
Encompass
The MCR Safety 6800 latex rubber-coated work gloves are designed for demanding work, offering excellent protection and enhanced gripping power. They feature a crinkle-textured rubber coating on the palm, fingers, and thumb, combined with canvas fabric and a duck cuff for lightweight and durable comfort.
  • Rubber coated canvas work gloves
  • Crinkle texture finish
  • Rubberized safety cuff
  • Interlock lining
  • Industrial grade for demanding tasks
  • Latex rubber coating for exceptional grip
  • Canvas glove with 3/4 dipped back
  • Wing thumb style
  • Suitable for automotive assembly, farming, and construction
Catalog No. 50-007-92048 Supplier MSC Supplier No. 83707232
The MCR Safety 6800 latex rubber-coated work gloves are designed for demanding work, offering excellent protection and enhanced gripping power. They feature a crinkle-textured rubber coating on the palm, fingers, and thumb, combined with canvas fabric and a duck cuff for lightweight and durable comfort.
  • Rubber coated canvas work gloves
  • Crinkle texture finish
  • Rubberized safety cuff
  • Interlock lining
  • Industrial grade for demanding tasks
  • Latex rubber coating for exceptional grip
  • Canvas glove with 3/4 dipped back
  • Wing thumb style
  • Suitable for automotive assembly, farming, and construction