Learn More
MSC MCR Safety Work Gloves, Medium, Latex-Coated, Acrylic, 1 Pair
Catalog No. 5000795898
Encompass
These work gloves are designed for general purpose use, featuring a latex coating and acrylic construction. They provide protection against abrasion, heat, cut, and puncture, making them suitable for various tasks.
  • Lime and blue color for high visibility
  • Acrylic lined for comfort
  • Crinkle grip for enhanced handling
  • Offers protection against abrasion, heat, cut, and puncture
Catalog No. 50-007-95898 Supplier MSC Supplier No. 02485324
These work gloves are designed for general purpose use, featuring a latex coating and acrylic construction. They provide protection against abrasion, heat, cut, and puncture, making them suitable for various tasks.
  • Lime and blue color for high visibility
  • Acrylic lined for comfort
  • Crinkle grip for enhanced handling
  • Offers protection against abrasion, heat, cut, and puncture