Learn More
MSC MCR Safety Work Gloves, Nitrile Coated, Nylon/Spandex, Medium, 1 ea
Catalog No. 5000803347
Encompass
MCR Safety Ninja® BNF gloves are designed for general purpose applications, featuring a 15 gauge nylon and spandex shell for superior comfort and dexterity. These gloves are coated with breathable nitrile foam, offering excellent wet and dry grip along with enhanced abrasion resistance. Sanitized to prevent odors and bacterial growth.
  • Features 15 gauge nylon / spandex shell
  • Coated palm and fingertips
  • Breathable nitrile foam (BNF) coating
  • Darker colors help hide dirt and grime
  • Ergonomic design minimizes hand fatigue
  • Provides excellent wet or dry grip
  • Offers great dexterity, flexibility, and movement
  • Sanitized to prevent odors and bacteria
  • Touchscreen compatible
  • OEKO-TEX certified
Catalog No. 50-008-03347 Supplier MSC Supplier No. 61562096
MCR Safety Ninja® BNF gloves are designed for general purpose applications, featuring a 15 gauge nylon and spandex shell for superior comfort and dexterity. These gloves are coated with breathable nitrile foam, offering excellent wet and dry grip along with enhanced abrasion resistance. Sanitized to prevent odors and bacterial growth.
  • Features 15 gauge nylon / spandex shell
  • Coated palm and fingertips
  • Breathable nitrile foam (BNF) coating
  • Darker colors help hide dirt and grime
  • Ergonomic design minimizes hand fatigue
  • Provides excellent wet or dry grip
  • Offers great dexterity, flexibility, and movement
  • Sanitized to prevent odors and bacteria
  • Touchscreen compatible
  • OEKO-TEX certified