Learn More
MSC MCR Safety Work Gloves, Nitrile-Coated Nylon, X-Large, 1 Pair
Catalog No. 5000766452
Encompass
MCR Safety N9674XL work gloves are designed for general purpose applications, featuring a nitrile coating for enhanced grip and protection, and a comfortable nylon lining.
  • Nitrile-coated for enhanced grip and protection
  • Nylon lined for comfort
  • Soft textured grip for improved handling
  • Designed for general purpose applications
  • Full fingered coverage
  • Provides abrasion and cut protection
  • Palm dipped with nitrile coating
  • X-large size
Catalog No. 50-007-66452 Supplier MSC Supplier No. 91609438
MCR Safety N9674XL work gloves are designed for general purpose applications, featuring a nitrile coating for enhanced grip and protection, and a comfortable nylon lining.
  • Nitrile-coated for enhanced grip and protection
  • Nylon lined for comfort
  • Soft textured grip for improved handling
  • Designed for general purpose applications
  • Full fingered coverage
  • Provides abrasion and cut protection
  • Palm dipped with nitrile coating
  • X-large size