Learn More
MSC MCR Safety Work Gloves, PVC Honeycomb Grip, Large, 1 Pair
Catalog No. 5000684874
Encompass
These string knit work gloves feature an uncured PVC honeycomb pattern for enhanced grip. Designed with a 7-gauge orange acrylic/polyester blended shell, they offer both breathability and comfort for general-purpose applications.
  • Breathable and comfortable
  • Color coded hem to identify sizes
  • Heavyweight 7-gauge acrylic/polyester blended shell
  • Provides excellent grip
  • Uncured PVC honeycomb pattern
  • Cut score A2
  • Abrasion score 3
  • Knit wrist cuff
Catalog No. 50-006-84874 Supplier MSC Supplier No. 52389996
These string knit work gloves feature an uncured PVC honeycomb pattern for enhanced grip. Designed with a 7-gauge orange acrylic/polyester blended shell, they offer both breathability and comfort for general-purpose applications.
  • Breathable and comfortable
  • Color coded hem to identify sizes
  • Heavyweight 7-gauge acrylic/polyester blended shell
  • Provides excellent grip
  • Uncured PVC honeycomb pattern
  • Cut score A2
  • Abrasion score 3
  • Knit wrist cuff