Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Long Length Natural Canvas Chore Gloves with Quilted Fleece Palm

Catalog No. 19058660 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Color:
Natural, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-058-660 Natural, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-058-660 Supplier MCR Safety Supplier No. 8516G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves feature a golden fleece palm, thumb, and forefinger with a rugged canvas material back. This multi purpose glove features a 5 inch plasticized gauntlet cuff for additional protection.

  • Regular weight golden chore gloves
  • Comfortable cotton polyester fabric
  • Gold fleece palm, index finger, and thumb
  • Canvas back
  • Clute pattern
  • Straight thumb
  • 5 inch plasticized gauntlet cuff

Specifications

Construction Jersey Knit
Cuff Material Plasticized
Color Natural, Yellow
Hand Specific
Material Cotton, Canvas
Product Type General Purpose Gloves
Size Large
Style Clute
For Use With (Application) General-purpose
Glove Cuff Color Natural
Glove Cuff Style Gauntlet
Hem Color Red
Packaging Type Polybag
Palm Type Gold Fleece
Thumb Style Straight
Length (English) 12.5 in.
Length (English) Cuff 4.5 in.
Length (Metric) 31.8 cm
Length (Metric) Cuff 11.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.