Learn More
Electron Microscopy Sciences Feather™ Stainless Steel Micro-Scalpel, Feather, 30 degree
SDP
Catalog No. 501899740
Encompass_Preferred
These disposable stainless steal MicroScalpels feature micro precision, ultra sharp points. They are sterilized by gamma radiation. Sure-grip aluminum handle that is color coded for easy angle identification. Comes in carbonate plastic case. Sure-grip aluminum handle-color coded for easy angle identification.
Angle: 30°
Catalog No. 50-189-9740 Supplier Electron Microscopy Sciences Supplier No. 7204530
These disposable stainless steal MicroScalpels feature micro precision, ultra sharp points. They are sterilized by gamma radiation. Sure-grip aluminum handle that is color coded for easy angle identification. Comes in carbonate plastic case. Sure-grip aluminum handle-color coded for easy angle identification.
Angle: 30°