Learn More
Research Products International Corp Micro-Tubes w/ Screw Cap, Brown, 2.0ml, Free Standing, 1000/cs
SDP
Catalog No. 502229771
Encompass
Dark brown color tubes block light transmission that can adversely affect fluorescent and other light sensitive samples. Polypropylene tubes withstand high speed centrifugation (up to 17,000xg), autoclaving, boiling and freezer storage. Screw cap with ''O'' ring provides superior sealing during sample processing. Tubes and caps are supplied in separate bags.
Ideal for light sensitive samples
Filter UV light
Leak Proof
RNAse, DNAse, DNA & pyrogen free
Catalog No. 50-222-9771 Supplier Research Products International Corp Supplier No. 144548SI
Dark brown color tubes block light transmission that can adversely affect fluorescent and other light sensitive samples. Polypropylene tubes withstand high speed centrifugation (up to 17,000xg), autoclaving, boiling and freezer storage. Screw cap with ''O'' ring provides superior sealing during sample processing. Tubes and caps are supplied in separate bags.
Ideal for light sensitive samples
Filter UV light
Leak Proof
RNAse, DNAse, DNA & pyrogen free