Learn More
Electron Microscopy Sciences Micromesh 4 Compartment Cassette, Fluo-Yellow
SDP
Catalog No. 501899229
Encompass_Preferred
Four compartments have 1676 square openings (0.38 mm) for improved fluid exchange. Made of acetyl polymer, resistant to most histological chemicals. Keeps specimen submerged. Use for labeling machines and tissue processors.
Color: Fluo-Yellow
Catalog No. 50-189-9229 Supplier Electron Microscopy Sciences Supplier No. 70075FY
Four compartments have 1676 square openings (0.38 mm) for improved fluid exchange. Made of acetyl polymer, resistant to most histological chemicals. Keeps specimen submerged. Use for labeling machines and tissue processors.
Color: Fluo-Yellow