Learn More
Ibidi USA µ-Plate 24 Well Black ID 14 mm, Uncoated - Uncoated Polymer Coverslip
SDP
Catalog No. NC2018832
Encompass
The ibidi µ-Plate 24 Well Black ID 14 mm allows you to perform high resolution microscopy in a standard multi-well format. The plate is made out of a black polymer to eliminate well-to-well crosstalk during fluorescence microscopy. Ø 14 mm wells, #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box, individually packaged.
This item is not returnable. View return policy
Catalog No. NC2018832 Supplier Ibidi USA Supplier No. 82421
The ibidi µ-Plate 24 Well Black ID 14 mm allows you to perform high resolution microscopy in a standard multi-well format. The plate is made out of a black polymer to eliminate well-to-well crosstalk during fluorescence microscopy. Ø 14 mm wells, #1.5 polymer coverslip, hydrophobic, sterilized, 15 pcs./box, individually packaged.