Learn More
World Precision Instrument Pre-Pulled Glass Pipette Samplers-TIPMIX05-10
SDP
Catalog No. 50821977
Encompass
WPI can quickly supply your need for consistently sized pre-pulled glass micropipettes. Tip diameters (ID) range from 0.1 to 10 µm. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the pipette and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass.
Catalog No. 50-821-977 Supplier World Precision Instrument Supplier No. TIPMIX0510
WPI can quickly supply your need for consistently sized pre-pulled glass micropipettes. Tip diameters (ID) range from 0.1 to 10 µm. Silanized Tips (Luer Shank) are available. Silanization waterproofs the glass to retard water when inserting into cell. This will not let the outside fluid run down the pipette and get inside so easily.These glass needles are fabricated from Schott Duran borosilicate glass.