Learn More
Electron Microscopy Sciences Microscope Slide Tray Folder Clips, Color: Assorted., 20/PK
SDP
Catalog No. 502090994
Encompass_Preferred
These Microscope Slide Tray folder Clips allows for the color coding of Microscope files as well as keeping the trays sealed shut so nothng falls out. The clips can be labeled with a permanenet marker or you can adhere labels. Perfect for histology, microscopy, and pathology labe, or in any research environment. Made for ABS and available in assortement of colors. Color: Assorted.
Catalog No. 50-209-0994 Supplier Electron Microscopy Sciences Supplier No. 7152501
These Microscope Slide Tray folder Clips allows for the color coding of Microscope files as well as keeping the trays sealed shut so nothng falls out. The clips can be labeled with a permanenet marker or you can adhere labels. Perfect for histology, microscopy, and pathology labe, or in any research environment. Made for ABS and available in assortement of colors. Color: Assorted.