Learn More
Electron Microscopy Sciences Visio-Tek™ Backlight Rack Microscope Stand
SDP
Catalog No. 501930540
Encompass_Preferred
Designed to be used with the complete line of Visio-tek digital microscopes. Up and down knobs are used to adjust the distance between microscope and object. With the removable X&Y crossed, you can slightly adjust the position of the object by rotating the knobs. You can turn on and off the LED backlight source thorough the brightness control knob to enhance the object clarity. Powered by 4 AA Batteries or USB.
Catalog No. 50-193-0540 Supplier Electron Microscopy Sciences Supplier No. 9950009
Designed to be used with the complete line of Visio-tek digital microscopes. Up and down knobs are used to adjust the distance between microscope and object. With the removable X&Y crossed, you can slightly adjust the position of the object by rotating the knobs. You can turn on and off the LED backlight source thorough the brightness control knob to enhance the object clarity. Powered by 4 AA Batteries or USB.