Learn More
Electron Microscopy Sciences ProScope EDU 1080P WIFI Microscope
SDP
Catalog No. 501929317
Encompass
The ProScope EDU WiFI Microscope features a super sharp 1080P (3MP) imager, 10X to 230X zoom, finite adjustable stand, apps for iOS and Android as well as software for Windows and Mac to capture stills and video. The ProScope EDU WiFi is perfect for all levels of science education and fits even the tightest budget. Includes 1080P Full HD WiFi Microscope, adjustable stand, USB cable, power adapter, software download, CD, calibration ruler, and 8GB Micro SD memory card.
Catalog No. 50-192-9317 Supplier Electron Microscopy Sciences Supplier No. 68350WIFI
The ProScope EDU WiFI Microscope features a super sharp 1080P (3MP) imager, 10X to 230X zoom, finite adjustable stand, apps for iOS and Android as well as software for Windows and Mac to capture stills and video. The ProScope EDU WiFi is perfect for all levels of science education and fits even the tightest budget. Includes 1080P Full HD WiFi Microscope, adjustable stand, USB cable, power adapter, software download, CD, calibration ruler, and 8GB Micro SD memory card.