Learn More
Electron Microscopy Sciences MicroSlicer DTK-1000N Fresh and Fixed Tissue Slicer
SDP
Catalog No. 501931224
Encompass_Preferred
MicroSlicers are the standard models for cutting live tissues. Combines high basic performance and economic efficiency. Manual operation of the elevating mechanism which controls section thickness.
Catalog No. 50-193-1224 Supplier Electron Microscopy Sciences Supplier No. 93304
Add to Cart
Add to Cart
Request a Quote
MicroSlicers are the standard models for cutting live tissues. Combines high basic performance and economic efficiency. Manual operation of the elevating mechanism which controls section thickness.