Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human M-CSF / CSF-1 Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;MCF,M-CSF,CSF-1,Lanimostim,MCSF,CSF1
Supplier:  ACROBiosystems MCFH82E6200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human M-CSF / CSF-1 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human M-CSF, His Tag, Avi Tag (MCF-H82E6) is expressed from human 293 cells (HEK293). It contains AA Glu 33 - Arg 255 (Accession # P09603-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.8 kDa. The protein migrates as 35-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF R, Fc Tag (Cat. No. CSR-H5258) at 2 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF, His Tag, Avi Tag (Cat. No. MCF-H82E6) with a linear range of 0.1-2 ng/mL (QC tested).
Catalog No. 50-165-7612