Learn More
ACROBiosystems Recombinant Protein;Human M-CSF R / CSF1R / CD115 Protein, Fc Tag, low endotoxin;HEK293;1MG;CSR,CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR,CSF-1R,CSF1 R
Catalog No. 501657954
Encompass_Preferred
Recombinant Protein;Human M-CSF R / CSF1R / CD115 Protein, Fc Tag, low endotoxin;1MG;Human M-CSF R, Fc Tag (CSR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # NP_005202.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 80.7 kDa. The protein migrates as 100-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 μg/mL (100 μL/well) can bind Human M-CSF R, Fc Tag (Cat. No. CSR-H5258) with a linear range of 0.1-4 ng/mL (QC tested).
Catalog No. 50-165-7954 Supplier ACROBiosystems Supplier No. CSRH52581MG
Recombinant Protein;Human M-CSF R / CSF1R / CD115 Protein, Fc Tag, low endotoxin;1MG;Human M-CSF R, Fc Tag (CSR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # NP_005202.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 80.7 kDa. The protein migrates as 100-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 μg/mL (100 μL/well) can bind Human M-CSF R, Fc Tag (Cat. No. CSR-H5258) with a linear range of 0.1-4 ng/mL (QC tested).