Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse M-CSF R / CSF1R / CD115 Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;CSR,CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR,CSF-1R,CSF1 R
Supplier:  ACROBiosystems CSRM82E825UG
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse M-CSF R / CSF1R / CD115 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Mouse M-CSF R, His Tag (CSR-M82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 (Accession # P09581-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 58.4 kDa. The protein migrates as 78-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 10 μg/mL (100 μL/well) can bind Biotinylated Mouse M-CSF R, His Tag (Cat. No. CSR-M82E8) with a linear range of 0.625-10 μg/mL (QC tested).
Catalog No. 50-165-8318