Learn More
Andwin Scientific MCT 400UL W/CAP 1000/PK
SDP
Catalog No. NC9691432
Encompass_Preferred
The Microcentrifuge Tubes With Caps by Evergreen Scientific are compatible with all standard microcentrifuges and cover a range from 250 to 2.0 mL. This product is constructed with polyethylene tubes that can withstand force to 6,000 g. The tubes are free of latex. It is sold in a pack of 1000.
Catalog No. NC9691432 Supplier Andwin Scientific Supplier No. 2143014020
The Microcentrifuge Tubes With Caps by Evergreen Scientific are compatible with all standard microcentrifuges and cover a range from 250 to 2.0 mL. This product is constructed with polyethylene tubes that can withstand force to 6,000 g. The tubes are free of latex. It is sold in a pack of 1000.