Learn More
Thomas Scientific MCT IND WRP 1.5ML ST 500PK
SDP
Catalog No. NC1870733
Encompass
Microcentrifuge Tubes, Sterile Microtubes (Individually wrapped), Volume: 1.5ml, Dimensions: 38 x 10mm Description: PP/gamma irradiated, 500/pk
Catalog No. NC1870733 Supplier Thomas Scientific Supplier No. 1158Q07
Microcentrifuge Tubes, Sterile Microtubes (Individually wrapped), Volume: 1.5ml, Dimensions: 38 x 10mm Description: PP/gamma irradiated, 500/pk