Learn More
Aviva Systems Biology Corp MCTP1 Antibody - middle region (ARP44440_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44440_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MCTP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MCTP1. Alternate names: FLJ22344ApplicationWB, IHC. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58808