Learn More
TARGETMOL CHEMICALS INC MCU-I4 25MG
SDP
Catalog No. 502576214
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MCU-i4 is a novel negative modulator of the MCU binding MICU1 and impairing muscle cell growth. purity: 99%
Catalog No. 50-257-6214 Supplier TARGETMOL CHEMICALS INC Supplier No. T903125MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MCU-i4 is a novel negative modulator of the MCU binding MICU1 and impairing muscle cell growth. purity: 99%