Learn More
TARGETMOL CHEMICALS INC MCUF-651 5MG
SDP
Catalog No. 502579272
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MCUF-651 is a guanylyl cyclase A receptor positive allosteric modulators EC50=0.45 uM. purity: 99%
Catalog No. 50-257-9272 Supplier TARGETMOL CHEMICALS INC Supplier No. T677345MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MCUF-651 is a guanylyl cyclase A receptor positive allosteric modulators EC50=0.45 uM. purity: 99%