Learn More
Enzo Life Sciences MD 2 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADICSA506E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the middle of human MD-2. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Purity: Ion exchange purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7185