Learn More
Medchemexpress LLC HY-134823 5mg , MD-222 CAS:2136246-72-3 Purity:>98%
SDP
Catalog No. 502261390
Encompass_Preferred
HY-134823 5mg MD-222 CAS: 2136246-72-3 MD-222 is the first-in-class highly potent PROTAC degrader of MDM2. MD-222 consists of ligands for Cereblon and MDM2. MD-222 induces rapid degradation of the MDM2 protein and activation of wild-type p53 in cells. MD-222 has anticancer effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1390 Supplier Medchemexpress LLC Supplier No. HY1348235MG
HY-134823 5mg MD-222 CAS: 2136246-72-3 MD-222 is the first-in-class highly potent PROTAC degrader of MDM2. MD-222 consists of ligands for Cereblon and MDM2. MD-222 induces rapid degradation of the MDM2 protein and activation of wild-type p53 in cells. MD-222 has anticancer effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.