Learn More
Medchemexpress LLC MD-224 100mg | 2136247-12-4 | 100 MG
SDP
Catalog No. 5000229143
Encompass_Preferred
MD-224 is a small-molecule proteolysis-targeting chimera (PROTAC) that selectively degrades MDM2 for research applications. It shows high cellular potency and is supplied as a solid or as a 10 mM solution in DMSO.
  • Selective MDM2 degrader for research use
  • High potency in cellular assays (subnanomolar activity)
  • Available as solid and 10 mM DMSO solution
  • High purity suitable for biochemical studies
  • Molecular formula C48H43Cl2FN6O6; molecular weight 889.80
Catalog No. 50-002-29143 Supplier Medchemexpress LLC Supplier No. HY114312100MG
MD-224 is a small-molecule proteolysis-targeting chimera (PROTAC) that selectively degrades MDM2 for research applications. It shows high cellular potency and is supplied as a solid or as a 10 mM solution in DMSO.
  • Selective MDM2 degrader for research use
  • High potency in cellular assays (subnanomolar activity)
  • Available as solid and 10 mM DMSO solution
  • High purity suitable for biochemical studies
  • Molecular formula C48H43Cl2FN6O6; molecular weight 889.80