Learn More
Medchemexpress LLC MD-224 10mM | 10MM 1ML
SDP
Catalog No. 5000229087
Encompass_Preferred
MD-224 is a first-in-class and highly potent small-molecule human murine double minute 2 (MDM2) degrader based on the proteolysistargeting chimera (PROTAC) concept MD-224 consists of ligands for Cereblon and MDM2 MD-224 induces rapid degradation of MDM2 at concentrations 1 nM in human leukemia cells and achieves an IC50 value of 1 5 nM in inhibition of growth of RS4 11 cells MD-224 has the potential to be a new class of anticancer agent[1] MD-224 is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-29087 Supplier Medchemexpress LLC Supplier No. HY11431210MM 1ML
MD-224 is a first-in-class and highly potent small-molecule human murine double minute 2 (MDM2) degrader based on the proteolysistargeting chimera (PROTAC) concept MD-224 consists of ligands for Cereblon and MDM2 MD-224 induces rapid degradation of MDM2 at concentrations 1 nM in human leukemia cells and achieves an IC50 value of 1 5 nM in inhibition of growth of RS4 11 cells MD-224 has the potential to be a new class of anticancer agent[1] MD-224 is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups