Learn More
Medchemexpress LLC MD-224 25mg | 2136247-12-4 | 25 MG
SDP
Catalog No. 5000308107
Encompass_Preferred
MD-224 is a proteolysis-targeting chimera (PROTAC) that potently degrades murine double minute 2 (MDM2). It is used as a research compound to probe the MDM2-p53 pathway and evaluate MDM2-targeted degradation in oncology and cell biology studies.
  • Potent MDM2 degrader active at sub-nanomolar concentrations
  • Suitable for cellular and biochemical studies of p53 signaling
  • Provided as a high-purity solid for research use
  • Available in laboratory-scale pack sizes, including 25 mg
  • Molecular formula C48H43Cl2FN6O6; molecular weight 889.80
Catalog No. 50-003-08107 Supplier Medchemexpress LLC Supplier No. HY11431225MG
MD-224 is a proteolysis-targeting chimera (PROTAC) that potently degrades murine double minute 2 (MDM2). It is used as a research compound to probe the MDM2-p53 pathway and evaluate MDM2-targeted degradation in oncology and cell biology studies.
  • Potent MDM2 degrader active at sub-nanomolar concentrations
  • Suitable for cellular and biochemical studies of p53 signaling
  • Provided as a high-purity solid for research use
  • Available in laboratory-scale pack sizes, including 25 mg
  • Molecular formula C48H43Cl2FN6O6; molecular weight 889.80