Learn More
Selleck Chemical LLC MD-224
SDP
Catalog No. 5000002771
Encompass_Preferred
MD-224 is a highly potent and effective degrader of the Human murine double minute 2 (MDM2) protein the main cellular inhibitor of the tumor suppressor p53 It strongly inhibits cell growth in leukemia cell lines with wild-type p53 and induces complete and lasting tumor regression in the RS4 11 xenograft tumor model
Catalog No. 50-000-02771 Supplier Selleck Chemical LLC Supplier No. S6900100MG
MD-224 is a highly potent and effective degrader of the Human murine double minute 2 (MDM2) protein the main cellular inhibitor of the tumor suppressor p53 It strongly inhibits cell growth in leukemia cell lines with wild-type p53 and induces complete and lasting tumor regression in the RS4 11 xenograft tumor model