Learn More
Medchemexpress LLC MD-224 500mg | 2136247-12-4 | 500 MG
SDP
Catalog No. 5000229738
Encompass_Preferred
MD-224 is a PROTAC small-molecule degrader of murine double minute 2 (MDM2) used in research to induce targeted protein degradation and study p53 pathway modulation. It is supplied as a high-purity solid with supporting analytical and safety documentation.
  • High purity suitable for cellular and in vivo research.
  • Contains an alkyne group for click chemistry conjugation.
  • Characterized by LC-MS and supplied with a certificate of analysis and SDS.
  • Available in multiple pack sizes and solution formats for flexible use.
  • Potent activity at sub-nanomolar concentrations for MDM2 degradation.
Catalog No. 50-002-29738 Supplier Medchemexpress LLC Supplier No. HY114312500MG
MD-224 is a PROTAC small-molecule degrader of murine double minute 2 (MDM2) used in research to induce targeted protein degradation and study p53 pathway modulation. It is supplied as a high-purity solid with supporting analytical and safety documentation.
  • High purity suitable for cellular and in vivo research.
  • Contains an alkyne group for click chemistry conjugation.
  • Characterized by LC-MS and supplied with a certificate of analysis and SDS.
  • Available in multiple pack sizes and solution formats for flexible use.
  • Potent activity at sub-nanomolar concentrations for MDM2 degradation.