Learn More
Medchemexpress LLC MD-224 50mg | 2136247-12-4 | 50 MG
SDP
Catalog No. 5000308106
Encompass_Preferred
MD-224 is a small-molecule proteolysis-targeting chimera (PROTAC) that induces degradation of the E3 ubiquitin ligase MDM2 and is used in biochemical and cell-based studies of the MDM2-p53 pathway.
  • Induces rapid MDM2 degradation at sub-nanomolar concentrations.
  • High purity suitable for research applications.
  • Molecular weight 889.80 g/mol; formula C48H43Cl2FN6O6.
  • Supplied as a 50 mg solid for laboratory use.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-003-08106 Supplier Medchemexpress LLC Supplier No. HY11431250MG
MD-224 is a small-molecule proteolysis-targeting chimera (PROTAC) that induces degradation of the E3 ubiquitin ligase MDM2 and is used in biochemical and cell-based studies of the MDM2-p53 pathway.
  • Induces rapid MDM2 degradation at sub-nanomolar concentrations.
  • High purity suitable for research applications.
  • Molecular weight 889.80 g/mol; formula C48H43Cl2FN6O6.
  • Supplied as a 50 mg solid for laboratory use.
  • Intended for research use only; not for human or clinical use.