Learn More
TARGETMOL CHEMICALS INC MD-224 5MG
SDP
Catalog No. 502581494
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MD-224 is a highly potent and efficacious MDM2 degrader based on the proteolysistargeting chimera (PROTAC) conceptand as a new class of anticancer agent. purity: 100%
Catalog No. 50-258-1494 Supplier TARGETMOL CHEMICALS INC Supplier No. T119805MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MD-224 is a highly potent and efficacious MDM2 degrader based on the proteolysistargeting chimera (PROTAC) conceptand as a new class of anticancer agent. purity: 100%