Learn More
Qualitest Usa MD-300S ELECTRONIC DENSIMETER
Supplier: Qualitest Usa MD300S
MD-300S Electronic Densimeter – For Solid & Liquid. Standard Configuration: Main Unit, Sensor (weighing tower), plastic water tank, support, holder, 200g calibration weight, thermometer, tweezers, AC adapter, steel angle, airtight windshield, RS-232C Interface. - Density resolution of 0.001g/cm3; - Capable of measuring specific gravity of liquid (other than those with high viscosity, which require optional parts) readily floating & pellets samples, such as urethane rubber, plastics, sponge, and wood; - Measures Solid & liquid; - Weight in the air; Weight in the water; Value of specific gravity; Volume; - Dimensions : (D) 218 x (W) 190 x (H) 170mm; - Weight :1.6 Kg.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.