Learn More
Selleck Chemical LLC MD2-IN-1
SDP
Supplier:  Selleck Chemical LLC S65732MG
Encompass_Preferred
MD2-IN-1 is an inhibitor of Myeloid differentiation protein 2 (MD2) with a KD value of 189 M
Catalog No. 50-000-02924